missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DcR2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16781-1-AP-150UL
This item is not returnable.
View return policy
Description
TRAIL (also known as Apo-2L) is a member of the tumor necrosis factor (TNF) ligand family that rapidly induces apoptosis in a variety of transformed cell lines. A fourth member of the emerging TRAIL receptor family, TRAIL-R4, has signaling capacity similar to that of TRAIL-R1 and TRAIL-R2 with respect to NF-kappaB activation, but differs in its inability to induce apoptosis. Like TRAIL-R1 and TRAIL-R2, this receptor shows widespread tissue expression. The human TRAIL-R4 gene has been mapped to chromosome 8p22-21, clustered with three other TRAIL receptors.Specifications
| DcR2 | |
| Polyclonal | |
| Unconjugated | |
| TNFRSF10D | |
| CD264, DCR2, Decoy receptor 2, TNFRSF10D, TRAIL R4, TRAIL receptor 4, TRAILR4, TRUNDD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8793 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.51 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UBN6 | |
| TNFRSF10D | |
| DcR2 Fusion Protein Ag10208 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction