missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DCPS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530566
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, HeLa, HepG2. Predicted reactivity: Human (99%), Mouse (87%), Rat (85%), Pig (87%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Necessary for the complete degradation of mRNAs, both in normal mRNA turnover and in nonsense-mediated mRNA decay. Removes the 7-methyl guanine cap structure from mRNA fragments shorter than 10 nucleotides that are produced by 3'->5' exosome-mediated mRNA decay. Releases m7GMP. Can also degrade m7GDP to m7GMP. Has no activity towards mRNA molecules longer than 25 nucleotides.
Specifications
| DCPS | |
| Polyclonal | |
| Unconjugated | |
| DCPS | |
| 5'-(N(7)-methyl 5'-triphosphoguanosine)-[mRNA] diphosphatase; ARS; Dcps; DCS1; DCS-1; decapping enzyme, scavenger; decapping scavenger enzyme; heat shock-like protein 1; HINT5; HINT-5; Hint-related 7meGMP-directed hydrolase; Histidine triad nucleotide-binding protein 5; histidine triad protein member 5; homolog of C. elegans 7meGMP-directed hydrolase dcs-1; HSL1; HSPC015; m7GpppX diphosphatase; mRNA decapping enzyme; scavenger mRNA-decapping enzyme DcpS | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 28960 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96C86 | |
| DCPS | |
| Recombinant protein encompassing a sequence within the center region of human DCPS. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction