missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCPS Antibody (OTI4H8), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71839IR
This item is not returnable.
View return policy
Description
DCPS Monoclonal specifically detects DCPS in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| DCPS | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DCPS | |
| Full length recombinant protein of human DCPS (NP_054745) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI4H8 | |
| Western Blot, Immunohistochemistry | |
| DCS1, DCS-1, decapping enzyme, scavenger, EC 3.-, heat shock-like protein 1, HINT5, HINT-5homolog of C. elegans 7meGMP-directed hydrolase dcs-1, Hint-related 7meGMP-directed hydrolase, Histidine triad protein member 5, HSL1, HSPC015, mRNA decapping enzyme, scavenger mRNA-decapping enzyme DcpS | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 28960.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction