missing translation for 'onlineSavingsMsg'
Learn More

DCPIB, MedChemExpress

Product Code. 16424876
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16424876

Brand: Medchem Express HY103371

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DCPIB is a selective, reversible and potent inhibitor of volume-regulated anion channels (VRAC).

DCPIB voltage-dependently activates potassium channels TREK1 and TRAAK, and inhibits TRESK, TASK1 and TASK3 (IC50s: 0.14, 0.95, 50.72 μM, respectively). DCPIB is also a selective blocker of swelling-induced chloride current (ICl,swell), with an IC50 of 4.1 μM. DCPIB is a useful tool for investigating structure-function studies of K2P channels.

Specifications

Chemical Name or Material DCPIB
CAS 82749-70-0
Color White
Purity Grade Notes Research
Molecular Formula C22H28Cl2O4
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (233.99 mM; Need ultrasonic)
SMILES O=C(O)CCCOC1=CC2=C(C(C(C3CCCC3)(CCCC)C2)=O)C(Cl)=C1Cl
Molecular Weight (g/mol) 427.36
Formula Weight 427.36
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.