missing translation for 'onlineSavingsMsg'
Learn More

DCPIB, MedChemExpress

Product Code. 16486557
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486557 5 mg 5mg
16460207 50 mg 50mg
16424876 100 mg 100mg
16423838 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16486557 Supplier Medchem Express Supplier No. HY103371
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress DCPIB is a selective, reversible and potent inhibitor of volume-regulated anion channels (VRAC).

DCPIB voltage-dependently activates potassium channels TREK1 and TRAAK, and inhibits TRESK, TASK1 and TASK3 (IC50s: 0.14, 0.95, 50.72 μM, respectively). DCPIB is also a selective blocker of swelling-induced chloride current (ICl,swell), with an IC50 of 4.1 μM. DCPIB is a useful tool for investigating structure-function studies of K2P channels.

Specifications

Chemical Name or Material DCPIB
CAS 82749-70-0
Color White
Purity Grade Notes Research
Molecular Formula C22H28Cl2O4
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (233.99 mM; Need ultrasonic)
SMILES O=C(O)CCCOC1=CC2=C(C(C(C3CCCC3)(CCCC)C2)=O)C(Cl)=C1Cl
Molecular Weight (g/mol) 427.36
Formula Weight 427.36
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.