missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DCP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534455
This item is not returnable.
View return policy
Description
A suggested positive control is 293 cell lysate. PA5-34455 can be used with blocking peptide PEP-1497.
The removal, or decapping, of eukaryotic mRNA is an important step in the degradation of mRNA. Decapping protein 2 (DCP2) is the major mRNA decapping enzyme in cells. It is a member of the Nudix hydrolases superfamily of proteins that predominantly catalyze the hydrolysis of small nucleoside diphosphate substrates linked to another moiety. DCP2 is widely expressed in multiple tissues at varying levels, with highest expression seen in testis and brain.
Specifications
| DCP2 | |
| Polyclonal | |
| Unconjugated | |
| DCP2 | |
| 2410015D23Rik; 5730537H01Rik; AL118268; DCP2; DCP2 decapping enzyme homolog; DCP2 decapping enzyme homolog (S. cerevisiae); decapping mRNA 2; Decapping protein 2; DFLJ33245; hDpc; m7GpppN-mRNA hydrolase; mRNA decapping enzyme 2; mRNA-decapping enzyme 2; Nucleoside diphosphate-linked moiety X motif 20; nudix (nucleoside diphosphate linked moiety X)-type motif 20; nudix (nucleoside diphosphate linked moiety X)-type motif 20 (NUDT20); Nudix motif 20; nudix-type motif 20; NUDT20; RGD1562909 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 167227, 291604, 70640 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8IU60, Q9CYC6 | |
| DCP2 | |
| An 18 amino acid synthetic peptide near the carboxy terminus of human DCP2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction