missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38521-100ul
This item is not returnable.
View return policy
Description
DCP2 Polyclonal antibody specifically detects DCP2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| DCP2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| DCP2 decapping enzyme homolog (S. cerevisiae), EC 3.-, FLJ33245, hDpc, mRNA-decapping enzyme 2, nudix (nucleoside diphosphate linked moiety X)-type motif 20, Nudix motif 20, NUDT20Nucleoside diphosphate-linked moiety X motif 20 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human DCP2 (NP_001229306.1).,, Sequence:, METKRVEIPGSVLDDLCSRFILHIPSEERDNAIRVCFQIELAHWFYLDFYMQNTPGLPQCGIRDFAKAVFSHCPFLLPQGEDVEKVLDEWKEYKMGVPTYGAIILDETLENVLLVQGYLAKSGWGFPKGKVNKEEAPHDCAAREVFEETGFDIKDYICKDDYIELRINDQLARLYIIPGIPKDTKFNPKTRREIRNIEWFSIEKLPCHRNDMTPKSKLGLAPNKFFMAIPFIRPLRDWLS | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 167227 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction