missing translation for 'onlineSavingsMsg'
Learn More

DCP2 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18400935 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400935 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400935 Supplier Novus Biologicals Supplier No. NBP141070AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DCP2 Polyclonal antibody specifically detects DCP2 in Human samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCP2
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DCP2 decapping enzyme homolog (S. cerevisiae), EC 3.-, FLJ33245, hDpc, mRNA-decapping enzyme 2, nudix (nucleoside diphosphate linked moiety X)-type motif 20, Nudix motif 20, NUDT20Nucleoside diphosphate-linked moiety X motif 20
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 370 and 420 of human mRNA-decapping enzyme 2 using the numbering given in entry NP_689837.2 (GeneID 167227).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 167227
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.