missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCP1B Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-04009APC
This item is not returnable.
View return policy
Description
DCP1B Polyclonal antibody specifically detects DCP1B in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)
Specifications
| DCP1B | |
| Polyclonal | |
| Immunoprecipitation, Western Blot (Negative) | |
| DCP1, DCP1 decapping enzyme homolog B (S. cerevisiae), decapping enzyme hDcp1b, EC 3.-, FLJ31638, hDcp1b, mRNA-decapping enzyme 1B | |
| Maps to a region between residue 250 and 300 of human DCP1 Decapping Enzyme Homolog B using the numbering given in entry NP_689853.3 (GeneID 196513). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 196513 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction