missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DCLK3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12809062
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12809062 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12809062 Supplier Invitrogen™ Supplier No. PA520911

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is K562 cell lysate. PA5-20911 can be used with blocking peptide PEP-1025.

DCLK3 is one of three doublecortin-like kinases similar to the Ca2+/calmodulin-dependent protein kinase (CaMK) family. DCLK3 mRNA, like that of the homologous DCLK1 and DCLK3, is highly expressed in adult brain, but only DCLK3 transcripts are present in liver and kidney, suggesting that DCLK3 may play other roles than in cortical development. The DCLK proteins are homologous to Doublecortin (DCX), a protein that is mutated in X-linked human lissencephaly. In mouse models where the DCX gene has been disrupted, DCLK1 expression increases slightly and appears to compensate for the loss of DCX, as mice mutant for both DCX and DCLK1 show a severe phenotype including perinatal lethality, disorganized neocortical layering, and profound hippocampal cytoarchitectural disorganization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCLK3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene DCLK3
Gene Accession No. Q8BWQ5, Q9C098
Gene Alias BC056929; C730036H08; CLICK-I and II-related; Click-I, -II related; CLICK-I,II-related; CLR; Dcamkl3; DCDC3C; DCK3; Dclk3; doublecortin and CaM kinase-like 3; doublecortin domain-containing protein 3C; doublecortin like kinase 3; doublecortin-like and CAM kinase-like 3; Doublecortin-like kinase 3; hCG_16695; KIAA1765; RGD1309232; serine/threonine-protein kinase DCLK3
Gene Symbols DCLK3
Host Species Rabbit
Immunogen A 14 amino acid peptide near the amino terminus of human DCLK3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245038, 85443
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.