missing translation for 'onlineSavingsMsg'
Learn More

DCLK3 Antibody, Biotin, Novus Biologicals™

Product Code. 18427770 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427770 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427770 Supplier Novus Biologicals Supplier No. NBP177129B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DCLK3 Polyclonal antibody specifically detects DCLK3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCLK3
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias CLICK-I, DCAMKL3, DCDC3CCLR, doublecortin and CaM kinase-like 3, Doublecortin domain-containing protein 3C, Doublecortin-like and CAM kinase-like 3, doublecortin-like kinase 3EC 2.7.11.1, EC 2.7.11, II-related, KIAA1765DCK3, serine/threonine-protein kinase DCLK3
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human DCLK3. The immunogen is located within the last 50 amino acids of DCLK3. Amino Acid Squence: SEGHFRSQHKRVVE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 85443
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.