missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCLK1 Rabbit anti-Human, Mouse, Rat, Clone: 8C8T3, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | DCLK1 |
|---|---|
| Clone | 8C8T3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18369096
|
Novus Biologicals
NBP3-16391-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18331517
|
Novus Biologicals
NBP3-16391-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DCLK1 Monoclonal antibody specifically detects DCLK1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| DCLK1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 9201 | |
| IgG | |
| Affinity purified |
| 8C8T3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DCAMKL1CLICK1, DCDC3ACL1, DCLK, doublecortin and CaM kinase-like 1, Doublecortin domain-containing protein 3A, doublecortin-like kinase 1Doublecortin-like and CAM kinase-like 1, EC 2.7.11, KIAA0369EC 2.7.11.1, serine/threonine-protein kinase DCLK1 | |
| A synthetic peptide corresponding to a sequence within amino acids 600-700 of human DCLK1 (O15075). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title