missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCLK1 Polyclonal Antibody

DCLK1 Polyclonal Antibody
Brand: Thermo Scientific PA520908
This item is not returnable.
View return policy
Description
DCLK1 Polyclonal Antibody for Western Blot, IHC
Specifications
| DCLK1 | |
| Polyclonal | |
| Unconjugated | |
| DCLK1 | |
| 1700113D08Rik; 2810480F11Rik; AI836758; Ania4; Calcium/calmodulin-dependent protein kinase type I-like CPG16; CL1; CLICK1; Click-I; CLICK-I beta; Cpg16; Dcamkl1; DCDC3A; Dcl; Dclk; Dclk1; double cortin and calcium/calmodulin-dependent protein kinase-like 1; doublecortin and calcium/calmodulin-dependent protein kinase-like 1; doublecortin domain-containing protein 3A; doublecortin like kinase 1; doublecortin-like and CAM kinase-like 1; Doublecortin-like kinase 1; KIAA0369; mKIAA0369; RP11-113P14.1; Serine/threonine-protein kinase DCLK1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13175, 83825, 9201 | |
| 4°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O08875, O15075, Q9JLM8 | |
| DCLK1 | |
| A 14 amino acid synthetic peptide near the amino terminus of human DCLK1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction