missing translation for 'onlineSavingsMsg'
Learn More

DCLK1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA595770

Product Code. 16558611

  • £439.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: VDVDQRFSAV QVLEHPWVND DGLPENEHQL SVAGKIKKHF NTGPKPNSTA AGVSVIALDH GFTIKRSGSL DYYQQPGMYW IRPPLLIRRG RFSDEDATRM; Positive Samples: HepG2, A-549, 22Rv1, Mouse heart

DCAMKL1 (DCLK1) serine/threonine kinase or DCLK1 or doublecortin-like kinase 1 contains two N-terminal doublecortin domains (which bind microtubules and regulate microtubule polymerization), a C-terminal serine/threonine protein kinase domain (which shows substantial homology to Ca2+/calmodulin-dependent protein kinase), and a serine/proline-rich domain in between the doublecortin and the protein kinase domains (which mediates multiple protein-protein interactions). DCAMKL1 is a microtubule-associated kinase that can undergo autophosphorylation. DCAMKL1 has microtubule-polymerizing activity that is independent of its protein kinase activity. DCAMKL1 is involved in several different cellular processes, including neuronal migration, retrograde transport, neuronal apoptosis and neurogenesis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DCLK1
Polyclonal
Unconjugated
DCLK1
1700113D08Rik, 2810480F11Rik, AI836758, Ania4, Calcium/calmodulin-dependent protein kinase type I-like CPG16, CL1, CLICK1, Click-I, CLICK-I beta, Cpg16, Dcamkl1, DCDC3A, Dcl, Dclk, Dclk1, double cortin and calcium/calmodulin-dependent protein kinase-like 1, doublecortin and calcium/calmodulin-dependent protein kinase-like 1, doublecortin domain-containing protein 3A, doublecortin like kinase 1, doublecortin-like and CAM kinase-like 1, Doublecortin-like kinase 1, KIAA0369, mKIAA0369, RP11-113P14.1, Serine/threonine-protein kinase DCLK1
Rabbit
Affinity Chromatography
RUO
13175, 9201
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
2.29 mg/mL
PBS with 50% glycerol and 0.01% thimerosal, pH 7.3
O15075, Q9JLM8
DCLK1
Recombinant fusion protein containing a sequence corresponding to amino acids 630-729 of human DCAMKL1/DCAMKL1/DCLK1 (NP_0047251)
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.