missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DCLK1 Monoclonal Antibody (C9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA550941
This item is not returnable.
View return policy
Description
DCAMKL1 (DCLK1) serine/threonine kinase or DCLK1 or doublecortin-like kinase 1 contains two N-terminal doublecortin domains (which bind microtubules and regulate microtubule polymerization), a C-terminal serine/threonine protein kinase domain (which shows substantial homology to Ca2+/calmodulin-dependent protein kinase), and a serine/proline-rich domain in between the doublecortin and the protein kinase domains (which mediates multiple protein-protein interactions). DCAMKL1 is a microtubule-associated kinase that can undergo autophosphorylation. DCAMKL1 has microtubule-polymerizing activity that is independent of its protein kinase activity. DCAMKL1 is involved in several different cellular processes, including neuronal migration, retrograde transport, neuronal apoptosis and neurogenesis.
Specifications
| DCLK1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| O08875, O15075 | |
| DCLK1 | |
| Recombinant Human DCLK1 protein, Tyr390-Val647 (Accession #O15075). | |
| 200 μL | |
| Primary | |
| Human, Pig, Rat | |
| Antibody | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| C9 | |
| Unconjugated | |
| DCLK1 | |
| 1700113D08Rik; 2810480F11Rik; AI836758; Ania4; Calcium/calmodulin-dependent protein kinase type I-like CPG16; CL1; CLICK1; Click-I; CLICK-I beta; Cpg16; Dcamkl1; DCDC3A; Dcl; Dclk; Dclk1; double cortin and calcium/calmodulin-dependent protein kinase-like 1; doublecortin and calcium/calmodulin-dependent protein kinase-like 1; doublecortin domain-containing protein 3A; doublecortin like kinase 1; doublecortin-like and CAM kinase-like 1; Doublecortin-like kinase 1; KIAA0369; mKIAA0369; RP11-113P14.1; Serine/threonine-protein kinase DCLK1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100623824, 83825, 9201 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction