missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DCLK1-IN-1

Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
DCLK1-IN-1 is a potent and selective doublecortin like kinase 1/2 (DCLK1/2) inhibitor (IC50 values are DCLK1 =9.5 and 57.2 nM and DCLK2 = 31 and 103 nM in binding and kinase assay, respectively). Exhibits no significant activity against ERK5, ACK, BRD4 and LRRK2. Exhibits DCLK inhibition in pancreatic ductal adenocarcinoma (PDAC) organoid models. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 5,11-Dihydro-2-[[2-methoxy-4-(4-methyl-1-piperazinyl)phenyl]amino]-11-methyl-5-(2,2,2-trifluoroethyl)-6 H-pyrimido[4,5-b][1,4 ]benzodiazepin-6-one |
| CAS | 2222635-15-4 |
| Quantity | 5 mg |
| Target | Other Kinase Inhibitors |
| Molecular Formula | C26H28F3N7O2 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction