missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCLK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£285.00 - £419.00
Specifications
| Antigen | DCLK1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18282104
|
Novus Biologicals
NBP2-58700 |
100 μL |
£419.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18669526
|
Novus Biologicals
NBP2-58700-25ul |
25 μL |
£285.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DCLK1 Polyclonal specifically detects DCLK1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| DCLK1 | |
| Polyclonal | |
| Rabbit | |
| Protein Kinase | |
| DCAMKL1CLICK1, DCDC3ACL1, DCLK, doublecortin and CaM kinase-like 1, Doublecortin domain-containing protein 3A, doublecortin-like kinase 1Doublecortin-like and CAM kinase-like 1, EC 2.7.11, KIAA0369EC 2.7.11.1, serine/threonine-protein kinase DCLK1 | |
| DCLK1 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 9201 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TSSVNGTPGSQLSTPRSGKSPSPSPTSPGSLRKQRSSQHGGSSTSLASTKVCSSMDENDGPGEEVSEEGFQIPATITERYKVGRTIGDGNFAVVKECVERSTAREYALKIIK | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title