missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DCL-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599615
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DCL-1.
CD302 is a C-type lectin receptor involved in cell adhesion and migration, as well as endocytosis and phagocytosis.
Specifications
| DCL-1 | |
| Polyclonal | |
| Unconjugated | |
| Cd302 | |
| 1110055L24Rik; AI159627; BIMLEC; CD302; CD302 antigen; CD302 molecule; CLEC13A; C-type lectin BIMLEC; C-type lectin domain family 13 member A; C-type lectin domain family 13, member A; DCL1; DCL-1; DEC205-associated C-type lectin 1; KIAA0022; RGD1307606; Type I transmembrane C-type lectin receptor DCL-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 66205, 9936 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8IX05, Q9DCG2 | |
| Cd302 | |
| A synthesized peptide derived from human CD302(Accession Q8IX05), corresponding to amino acid residues C56-D106. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction