missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DCL-1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17770464
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17770464 200 μL 200µL
17773434 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17770464 Supplier Invitrogen™ Supplier No. PA5119055

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DCL-1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

CD302 is a C-type lectin receptor involved in cell adhesion and migration, as well as endocytosis and phagocytosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCL-1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Cd302
Gene Accession No. Q9DCG2
Gene Alias 1110055L24Rik; AI159627; BIMLEC; CD302; CD302 antigen; CD302 molecule; CLEC13A; C-type lectin BIMLEC; C-type lectin domain family 13 member A; C-type lectin domain family 13, member A; DCL1; DCL-1; DEC205-associated C-type lectin 1; KIAA0022; RGD1307606; Type I transmembrane C-type lectin receptor DCL-1
Gene Symbols Cd302
Host Species Rabbit
Immunogen Recombinant protein C-Type Lectin Domain Family 13, Member A. The antigen corresponds to amino acid range 21-225 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66205
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.