missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DCL-1 Monoclonal Antibody (6B1F6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548676
This item is not returnable.
View return policy
Description
DCL-1 Monoclonal Antibody for Western Blot, ELISA
CD302 is a C-type lectin receptor involved in cell adhesion and migration, as well as endocytosis and phagocytosis.
Specifications
| DCL-1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q8IX05 | |
| Cd302 | |
| Purified recombinant fragment of human CD302 (AA: extra 23-168) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot, Western Blot | |
| 6B1F6 | |
| Unconjugated | |
| Cd302 | |
| 1110055L24Rik; AI159627; BIMLEC; CD302; CD302 antigen; CD302 molecule; CLEC13A; C-type lectin BIMLEC; C-type lectin domain family 13 member A; C-type lectin domain family 13, member A; DCL1; DCL-1; DEC205-associated C-type lectin 1; KIAA0022; RGD1307606; Type I transmembrane C-type lectin receptor DCL-1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 9936 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction