missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DCK Monoclonal Antibody (GT7710)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527741
This item is not returnable.
View return policy
Description
Positive Control: HeLa, HepG2, 293T, A549, H1299, HCT116 Predicted Reactivity: Rhesus Monkey (96%), Bovine (95%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Deoxycytidine kinase (DCK) is required for the phosphorylation of several deoxyribonucleosides and their nucleoside analogs. Deficiency of DCK is associated with resistance to antiviral and anticancer chemotherapeutic agents. Conversely, increased deoxycytidine kinase activity is associated with increased activation of these compounds to cytotoxic nucleoside triphosphate derivatives. DCK is clinically important because of its relationship to drug resistance and sensitivity.
Specifications
| DCK | |
| Monoclonal | |
| 1.33 mg/mL | |
| PBS with no preservative | |
| P27707, P43346, P48769 | |
| DCK | |
| Recombinant protein encompassing a sequence within the center region of human DCK. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| GT7710 | |
| Unconjugated | |
| DCK | |
| DCK; Deoxycytidine kinase; deoxynucleoside kinase | |
| Mouse | |
| Protein G | |
| RUO | |
| 13178, 1633, 79127 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction