missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCIR/CLEC4A Antibody (9E8) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
£196.00 - £548.00
Specifications
| Antigen | DCIR/CLEC4A |
|---|---|
| Clone | 9E8 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18614077
|
Novus Biologicals
NBP3-09023-0.2mg |
0.2 mg |
£548.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633788
|
Novus Biologicals
NBP3-09023-0.025mg |
0.025 mg |
£196.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DCIR/CLEC4A Monoclonal antibody specifically detects DCIR/CLEC4A in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| DCIR/CLEC4A | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor | |
| This antibody was raised by immunising BALB/c mice with a dendritic cell immunoreceptor ectodomain-human Fc fusion protein. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 9E8 | |
| Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Immunology, Innate Immunity | |
| PBS | |
| 50856 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title