missing translation for 'onlineSavingsMsg'
Learn More

DCI Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£145.00 - £417.00

Specifications

Antigen DCI
Dilution Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30231999
View Documents
Novus Biologicals
NBP3-35218-20ul
20 μL
£145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30228282
View Documents
Novus Biologicals
NBP3-35218-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DCI Polyclonal antibody specifically detects DCI in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications

Specifications

DCI
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Unconjugated
Rabbit
metabolism
PBS (pH 7.3), 50% glycerol
1632
IgG
Affinity purified
Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Polyclonal
Purified
RUO
Human, Mouse
acetylene-allene isomerase, D2-enoyl-CoA isomerase, D3, DCI3,2-trans-enoyl-CoA isomerase3,2 trans-enoyl-CoA isomerase, Delta(2)-enoyl-CoA isomerase, Delta(3), delta3, delta2-enoyl-CoA isomerase, dodecenoyl-CoA delta isomerase (3,2 trans-enoyl-CoA isomerase), dodecenoyl-CoA isomerase3,2-trans-enoyl-CoA isomerase, mitochondrial, dodecenoyl-Coenzyme A delta isomerase (3,2 trans-enoyl-Coenzyme A isomerase), EC 5.3.3.8, enoyl-CoA delta isomerase 13,2 trans-enoyl-Coenzyme A isomerase
Recombinant fusion protein containing a sequence corresponding to amino acids 63-302 of human DCI (NP_001910.2).,, Sequence:, KNPPVNSLSLEFLTELVISLEKLENDKSFRGVILTSDRPGVFSAGLDLTEMCGRSPAHYAGYWKAVQELWLRLYQSNLVLVSAINGACPAGGCLVALTCDYRILADNPRYCIGLNETQLGIIAPFWLKDTLENTIGHRAAERALQLGLLFPPAEALQVGIVDQVVPEEQVQSTALSAIAQWMAIPDHARQLTKAMMRKATASRLVTQRDADVQNFVSFISKDSIQKSLQMYLERLKEEKG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documenti

Documenti

Product Certifications
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato