Learn More
DCHS1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA553683
Description
Immunogen sequence: HSSGRGSAEA AEDDEIRMIN EFPRVASVAS SLAARGPDSG IQQDADGLSD TSCEPPAPDT WYKGRKAGLL LPGAGATLYR EEGPPATATA FLGGCGLSPA PTGDYGFPAD GKPCVAGALT AIVAGEEELR GSYNWDYLLS WCPQFQPLAS Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 100%.
DCHS1 Polyclonal antibody specifically detects DCHS1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Specifications
| DCHS1 | |
| Polyclonal | |
| Unconjugated | |
| DCHS1 | |
| Cadherin-19; cadherin-25; cadherin-related family member 6; CDH19; CDH25; CDHR6; dachsous cadherin-related 1; DCHS1; FIB1; fibroblast cadherin 1; fibroblast cadherin FIB1; Fibroblast cadherin-1; KIAA1773; MVP2; PCDH16; Protein dachsous homolog 1; protocadherin-16; VMLDS1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8642 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry | |
| 0.9 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96JQ0 | |
| DCHS1 | |
| Recombinant protein corresponding to Human DCHS1 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only