missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DCG IV
GREENER_CHOICE

Product Code. p-4783397
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
This item is not returnable. View return policy

Product Code. 18094262

Brand: Tocris Bioscience™ 0975/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly potent group II mGlu agonist; also NMDA agonist

DCG IV is a presynaptic depressant; highly potent agonist for group II mGlu receptors.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2 S,2 'R,3 'R)-2-(2 ',3 '-Dicarboxycyclopropyl)glycine
CAS 147782-19-2
Assay Percent Range >98%
Quantity 50 mg
Solubility Soluble to 100mM in water
Target Group II mGluR Agonists
Molecular Formula C7H9NO6
Formula Weight Observed MW: 203.15
Purity 98%

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.