missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DCDC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583198
This item is not returnable.
View return policy
Description
Immunogen sequence: LEGKLVESGA ELENGQFYVA VGRDKFKKLP YSELLFDKST MRRPFGQKAS SLPPIVGSRK SKGSGNDRHS KSTVGSSDNS SPQPL Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 92%.
This gene encodes a member of the doublecortin family. The protein encoded by this gene contains two doublecortin domains. The doublecortin domain has been demonstrated to bind tubulin and enhance microtubule polymerization. Mutations in this gene have been associated with Reading Disability, also referred to as developmental dyslexia. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| DCDC2 | |
| Polyclonal | |
| Unconjugated | |
| DCDC2 | |
| AW492955; Dcdc2; Dcdc2a; DFNB66; doublecortin domain containing 2; doublecortin domain containing 2a; doublecortin domain-containing protein 2; HGNC:18141; Kiaa1154; MGC54817; NPHP19; Protein RU2S; RU2; RU2S | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51473 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UHG0 | |
| DCDC2 | |
| Recombinant protein corresponding to Human DCDC2. Recombinant protein control fragment (Product #RP-104637). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction