missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DCDC2 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA518592
This item is not returnable.
View return policy
Description
This antibody is predicted to react with human and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
This gene encodes a member of the doublecortin family. The protein encoded by this gene contains two doublecortin domains. The doublecortin domain has been demonstrated to bind tubulin and enhance microtubule polymerization. Mutations in this gene have been associated with Reading Disability, also referred to as developmental dyslexia. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| DCDC2 | |
| Polyclonal | |
| Unconjugated | |
| Dcdc2a | |
| AW492955; Dcdc2; Dcdc2a; DFNB66; doublecortin domain containing 2; doublecortin domain containing 2a; doublecortin domain-containing protein 2; HGNC:18141; Kiaa1154; MGC54817; NPHP19; Protein RU2S; RU2; RU2S | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 195208 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q5DU00 | |
| Dcdc2a | |
| Synthetic peptide sequence (PQPLKRKGKKDSN) corresponding to the internal amino acids of Dcdc2a (aa 271-284). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction