missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DCC Polyclonal Antibody
GREENER_CHOICE

Product Code. 15904715
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15904715 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15904715 Supplier Invitrogen™ Supplier No. PA579139

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue, Mouse Brain Tissue, U87 Cell, SW620 Cell, COLO320 Cell, 293T Cell, HELA Cell. IHC: Human Intestinal Cancer Tissue.

This gene encodes a netrin 1 receptor. The transmembrane protein is a member of the immunoglobulin superfamily of cell adhesion molecules, and mediates axon guidance of neuronal growth cones towards sources of netrin 1 ligand. The cytoplasmic tail interacts with the tyrosine kinases Src and focal adhesion kinase (FAK, also known as PTK2) to mediate axon attraction. The protein partially localizes to lipid rafts, and induces apoptosis in the absence of ligand. The protein functions as a tumor suppressor, and is frequently mutated or downregulated in colorectal cancer and esophageal carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene DCC
Gene Accession No. P43146, P70211, Q63155
Gene Alias C030036D22Rik; Colorectal cancer suppressor; colorectal tumor suppressor; CRC18; CRCR1; Dcc; DCC netrin 1 receptor; deleted in colorectal cancer protein; deleted in colorectal carcinoma; IGDCC1; Immunoglobulin superfamily DCC subclass member 1; immunoglobulin superfamily, DCC subclass, member 1; MRMV1; Netrin receptor DCC; NTN1R1; putative colorectal tumor suppressor; tumor suppressor protein DCC
Gene Symbols DCC
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of human DCC (162-178aa EVIGEPMPTIHWQKNQQ).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13176, 1630, 25311
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.