missing translation for 'onlineSavingsMsg'
Learn More

DCC Antibody (OTI4A1) [DyLight 405], Novus Biologicals™

Product Code. 30688819 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30688819 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688819 Supplier Novus Biologicals Supplier No. NBP344682V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DCC Monoclonal antibody specifically detects DCC in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCC
Applications Western Blot
Classification Monoclonal
Clone OTI4A1
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias Colorectal cancer suppressor, CRC18, CRCR1, deleted in colorectal cancer protein, deleted in colorectal carcinoma, IGDCC1colorectal tumor suppressor, Immunoglobulin superfamily DCC subclass member 1, immunoglobulin superfamily, DCC subclass, member 1, netrin receptor DCC, Tumor suppressor protein DCC
Host Species Mouse
Immunogen Full length human recombinant protein of human DCC (NP_005206) produced in HEK293T cell.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 1630
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.