missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCBLD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13168-1-AP-20UL
This item is not returnable.
View return policy
Description
This protein is a key enzyme in the catabolic pathway of odd-chain fatty acids, isoleucine, threonine, methionine, and valine. It is probably a dodecamer composed of six biotin-containing alpha subunits and six beta subunits. The protein is located in the mitochondrial matrix. Defects in PCCA are the cause o fpropionic acidemia type I (PA-1) [MIM:606054]. PA-1 is a life-threatening disease characterized by episodic vomiting, lethargy and ketosis, neutropenia, periodic thrombocytopenia, hypogammaglobulinemia, develop-mental retardation, and intolerance to protein.Specifications
| DCBLD2 | |
| Polyclonal | |
| Unconjugated | |
| DCBLD2 | |
| CLCP1, DCBLD2, ESDN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 131566, 155696, 73379 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q91ZV2, Q91ZV3, Q96PD2 | |
| Dcbld2 | |
| DCBLD2 Fusion Protein Ag3821 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction