missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DCBLD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA14270
This item is not returnable.
View return policy
Description
PA1-4270 detects ESDN from human, mouse and rat samples. PA1-4270 has been successfully used in Western blot procedures. PA1-4270 immunizing peptide corresponds to amino acid residues 399-416 from mouse and rat ESDN protein.
This protein is a key enzyme in the catabolic pathway of odd-chain fatty acids, isoleucine, threonine, methionine, and valine. It is probably a dodecamer composed of six biotin-containing alpha subunits and six beta subunits. The protein is located in the mitochondrial matrix. Defects in PCCA are the cause o fpropionic acidemia type I (PA-1) [MIM:606054]. PA-1 is a life-threatening disease characterized by episodic vomiting, lethargy and ketosis, neutropenia, periodic thrombocytopenia, hypogammaglobulinemia, develop-mental retardation, and intolerance to protein.
Specifications
| DCBLD2 | |
| Polyclonal | |
| Unconjugated | |
| DCBLD2 | |
| 1700055P21Rik; AW146002; CLCP1; coagulation factor V/VIII-homology domains protein 1; CUB, LCCL and coagulation factor V/VIII-homology domains protein 1; DCBLD2; discoidin; discoidin, CUB and LCCL domain containing 2; discoidin, CUB and LCCL domain-containing protein 2; Endothelial and smooth muscle cell-derived neuropilin-like protein; ESDN | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 131566, 155696, 73379 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q91ZV2, Q91ZV3, Q96PD2 | |
| DCBLD2 | |
| Synthetic Peptide: Q(399) D K I F Q G N K D Y H K D V R N N(416). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction