missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DCBLD2/ESDN Antibody (FA19-1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | DCBLD2/ESDN |
|---|---|
| Clone | FA19-1 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30046782
|
Novus Biologicals™
NBP3-28795-100ug |
100 μg |
£247.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046300
|
Novus Biologicals™
NBP3-28795-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30045399
|
Novus Biologicals™
NBP3-28795-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DCBLD2/ESDN Monoclonal antibody specifically detects DCBLD2/ESDN in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| DCBLD2/ESDN | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Signal Transduction | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CLCP11700055P21Rik, CUB, LCCL and coagulation factor V/VIII-homology domains protein 1, discoidin, CUB and LCCL domain containing 2, discoidin, CUB and LCCL domain-containing protein 2, Endothelial and smooth muscle cell-derived neuropilin-like protein, ESDNcoagulation factor V/VIII-homology domains protein 1 | |
| DCBLD2/ ESDN | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| FA19-1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q96PD2 | |
| 131566 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title