missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DCBLD2/ESDN Polyclonal specifically detects DCBLD2/ESDN in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | DCBLD2/ESDN |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Formulation | PBS |
| Gene Alias | CLCP11700055P21Rik, CUB, LCCL and coagulation factor V/VIII-homology domains protein 1, discoidin, CUB and LCCL domain containing 2, discoidin, CUB and LCCL domain-containing protein 2, Endothelial and smooth muscle cell-derived neuropilin-like protein, ESDNcoagulation factor V/VIII-homology domains protein 1 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant DCBLD2/ESDN (NP_563615.3; Met1-Ala482). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?