Learn More
DCBLD2/ESDN Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93567APC
This item is not returnable.
View return policy
Description
DCBLD2/ESDN Polyclonal antibody specifically detects DCBLD2/ESDN in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Polyclonal | |
| APC | |
| Store at 4C in the dark. | |
| Purified |