missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCAR/CLEC4B Antibody (349214), DyLight 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2757M
This item is not returnable.
View return policy
Description
DCAR/CLEC4B Monoclonal antibody specifically detects DCAR/CLEC4B in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| DCAR/CLEC4B | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6320 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 349214 | |
| Flow Cytometry, CyTOF-ready | |
| CLECSF3, LSLCL, P47, SCGF | |
| Mouse myeloma cell line NS0-derived recombinant mouse DCAR/CLEC4B, Gln42-Leu209, Accession # AAR13070 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction