missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DCAMKL1 (DCLK1), GST Tag Recombinant Protein

Product Code. 15910283
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
This item is not returnable. View return policy

Product Code. 15910283

Brand: Invitrogen™ A30495

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This DCAMKL1 (DCLK1) recombinant human protein (full length) was expressed in insect cells. DCAMKL1 (DCLK1) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for DCAMKL1 (DCLK1) is NP_004725.1.

DCAMKL1 (DCLK1) serine/threonine kinase or DCLK1 or doublecortin-like kinase 1 contains two N-terminal doublecortin domains (which bind microtubules and regulate microtubule polymerization), a C-terminal serine/threonine protein kinase domain (which shows substantial homology to Ca2+/calmodulin-dependent protein kinase), and a serine/proline-rich domain in between the doublecortin and the protein kinase domains (which mediates multiple protein-protein interactions). DCAMKL1 is a microtubule-associated kinase that can undergo autophosphorylation. DCAMKL1 has microtubule-polymerizing activity that is independent of its protein kinase activity. DCAMKL1 is involved in several different cellular processes, including neuronal migration, retrograde transport, neuronal apoptosis and neurogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O15075
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 0.1mM EDTA, 25% glycerol, 0.1mM PMSF, 150mM NaCl, 0.25mM DTT, 10mM glutathione and no preservative; pH 7.5
Gene ID (Entrez) 9201
Molecular Weight (g/mol) 107.4 kDa
Name Human DCAMKL1 (DCLK1), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 1700113D08Rik; 2810480F11Rik; AI836758; Ania4; Calcium/calmodulin-dependent protein kinase type I-like CPG16; CL1; CLICK1; Click-I; CLICK-I beta; Cpg16; Dcamkl1; DCDC3A; Dcl; Dclk; Dclk1; double cortin and calcium/calmodulin-dependent protein kinase-like 1; doublecortin and calcium/calmodulin-dependent protein kinase-like 1; doublecortin domain-containing protein 3A; doublecortin like kinase 1; doublecortin-like and CAM kinase-like 1; Doublecortin-like kinase 1; KIAA0369; mKIAA0369; RP11-113P14.1; Serine/threonine-protein kinase DCLK1
Common Name DCLK1
Gene Symbol DCLK1
KinaseFamily DCAMKL Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Full Length, Recombinant, Active
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.