missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DCAF7 Recombinant Rabbit Monoclonal Antibody (23GB3115), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£162.00 - £387.00

Specifications

Antigen DCAF7
Clone 23GB3115
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30088025
GREENER_CHOICE
View Documents
Invitrogen™
MA552594
100 μL
£387.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30087666
View Documents
Invitrogen™
MA552595
20 μL
£162.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DCAF7 Recombinant Monoclonal antibody specifically detects DCAF7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications

Specifications

DCAF7
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse, Rat
P61962, P61963
10238, 303602, 71833
IgG
Affinity chromatography
Antibody
23GB3115
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
1700012F10Rik; 2610037L01Rik; AN11; AN11 homolog; C86529; DCAF7; DDB1 and CUL4 associated factor 7; DDB1- and CUL4-associated factor 7; dirty south; dys; fj83g06; HAN11; han11l; human anthocyanin; RGD1305140; seven-WD-repeat protein of the AN11 family-1; SWAN-1; Unknown (protein for MGC:134591); WD repeat domain 68; WD repeat-containing protein 68; WD repeat-containing protein An11 homolog; wdr68; WD-repeat protein; wu:fj83g06; zgc:63940
A synthesized peptide derived from DCAF7 (210-248AA).
Primary
-20°C
DCAF7
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.