missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCAF4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92192-0.1ml
This item is not returnable.
View return policy
Description
DCAF4 Polyclonal antibody specifically detects DCAF4 in Human samples. It is validated for Western Blot
Specifications
| DCAF4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DDB1 and CUL4 associated factor 4, DDB1- and CUL4-associated factor 4, DKFZp434K114, MGC20547, MGC46524, WD repeat domain 21, WD repeat domain 21A, WD repeat-containing protein 21A, WDR21, WDR21A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human DCAF4 (NP_056419.2). MNKSRWQSRRRHGRRSHQQNPWFRLRDSEDRSDSRAAQPAHDSGHGDDESPSTSSGTAGTSSVPELPGFYFDPEKKRYFRLLPGHNNCNPLTKESIRQKEMESKRLRLLQEEDRRKKIAR | |
| 0.1 mL | |
| Cell Biology | |
| 26094 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction