missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCAF12 Antibody, PerCP, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP1-77373PCP
This item is not returnable.
View return policy
Description
DCAF12 Polyclonal antibody specifically detects DCAF12 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| DCAF12 | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| DDB1- and CUL4-associated factor 12, CT102, DDB1 and CUL4 associated factor 12, KIAA1892, TCC52, WDR40A | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human TCC52. The immunogen is located within amino acids 390 - 440 of TCC52. Amino Acid Squence: LNHDETWRNYFSDID | |
| 0.1 mL | |
| Cell Biology | |
| 25853 | |
| Store at 4C in the dark. | |
| IgY |
| Western Blot, ELISA, Immunocytochemistry | |
| PerCP | |
| PBS | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction