missing translation for 'onlineSavingsMsg'
Learn More

DCAF12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£386.00

Specifications

Antigen DCAF12
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18255213
View Documents
Novus Biologicals
NBP1-56584
100 μL
£386.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DCAF12 Polyclonal specifically detects DCAF12 in Human samples. It is validated for Western Blot.
Specifications

Specifications

DCAF12
Polyclonal
Rabbit
Q5T6F0
25853
Synthetic peptides corresponding to WDR40A(WD repeat domain 40A) The peptide sequence was selected from the middle region of WDR40A. Peptide sequence TKSDARHNVSRVPVYAHITHKALKDIPKEDTNPDNCKVRALAFNNKNKEL.
Primary
Western Blot
Unconjugated
RUO
DDB1- and CUL4-associated factor 12, CT102, DDB1 and CUL4 associated factor 12, KIAA1892, TCC52, WDR40A
DCAF12
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.