missing translation for 'onlineSavingsMsg'
Learn More

DC8 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18373444 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18373444 20 μL 20µL
18351027 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18373444 Supplier Novus Biologicals Supplier No. NBP30342220UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DC8 Polyclonal specifically detects DC8 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DC8
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias C1orf48, chromosome 1 open reading frame 48, DC8, DKFZP566O1646, kinetochore-associated protein NSL1 homolog, MIS14, NSL1, MIND kinetochore complex component, homolog (S. cerevisiae)
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-281 of human DC8 (NP_056286.3).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 25936
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.