missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DC8 Polyclonal antibody specifically detects DC8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | DC8 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 150 and 200 of human DC8 (C1orf48) using the numbering given in entry NP_056286.2 (GeneID 25936 ). |
| Quantity | 0.1 mL |
| Primary or Secondary | Primary |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?