missing translation for 'onlineSavingsMsg'
Learn More

DC661, MedChemExpress

Product Code. 16495366
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495366 100 mg 100mg
16484498 10 mg 10mg
16480136 2 mg 2mg
16433116 25 mg 25mg
16407227 5 mg 5mg
16400817 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16495366 Supplier Medchem Express Supplier No. HY111621

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress DC661 is a potent palmitoyl-protein thioesterase 1 (PPT1) inhibitor, inhibits autophagy, and acts as an anti-lysosomal agent. Anti-cancer activity.

DC661 is a potent palmitoyl-protein thioesterase 1 (PPT1) inhibitor, inhibits autophagy, and acts as an anti-lysosomal agent. Anti-cancer activity.

Specifications

Chemical Name or Material DC661
CAS 1872387-43-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C31H39Cl2N5
Quantity 100 mg
Solubility Information DMSO : 62.5 mg/mL (113.11 mM; Need ultrasonic)
SMILES CN(CCCCCCNC1=CC=NC2=CC(Cl)=CC=C12)CCCCCCNC3=CC=NC4=CC(Cl)=CC=C34
Molecular Weight (g/mol) 552.58
Formula Weight 552.58
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.