missing translation for 'onlineSavingsMsg'
Learn More

DC1SMe, MedChemExpress

Product Code. 16464728
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464728 10 mg 10mg
16485546 100 mg 100mg
16427447 5 mg 5mg
16423226 25 mg 25mg
16421017 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16464728 Supplier Medchem Express Supplier No. HY112898

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DC1Sme, a DC1 derivative, exhibits IC50 values of 22 pM, 10 pM, 32 pM and 250 pM for Ramos, Namalwa, HL60/s and COLO 205 cancer cells, respectively. DC1, an analogue of the minor groove-binding DNA alkylator CC-1065, is a ADC Cytotoxin. DC1 can be used in synthesis of antibody-drug conjugates for the targeted treatment of cancer.

DC1Sme, a DC1 derivative, exhibits IC50 values of 22 pM, 10 pM, 32 pM and 250 pM for Ramos, Namalwa, HL60/s and COLO 205 cancer cells, respectively. DC1, an analogue of the minor groove-binding DNA alkylator CC-1065, is a ADC Cytotoxin. DC1 can be used in synthesis of antibody-drug conjugates for the targeted treatment of cancer.

Specifications

Chemical Name or Material DC1SMe
CAS 501666-85-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C35H30ClN5O4S2
Quantity 10 mg
SMILES O=C(C(N1)=CC2=C1C=CC(NC(CCSSC)=O)=C2)NC3=CC4=C(NC(C(N5C[C@@H](CCl)C6=C5C=C(O)C7=CC=CC=C67)=O)=C4)C=C3
Molecular Weight (g/mol) 684.23
Formula Weight 684.23
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.