missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ DC-STAMP/TM7SF4 Antibody (U.Rochester patent anti-DC-STAMP) - Humanized, Novus Biologicals™

Human Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-28686-50ug

 View more versions of this product

Product Code. 30044774

  • £162.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DC-STAMP/TM7SF4 Monoclonal antibody specifically detects DC-STAMP/TM7SF4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DC-STAMP/TM7SF4
Monoclonal
Unconjugated
Q9H295
Human
Protein A purified
RUO
81501
Human
Purified
Flow Cytometry, ELISA, Functional Assay
U.Rochester patent anti-DC-STAMP
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
dendritic cell-specific transmembrane protein, transmembrane 7 superfamily member 4
DCSTAMP
50 μg
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.