missing translation for 'onlineSavingsMsg'
Learn More

DC-SIGNR/CD299/CLEC4M Antibody (MM0241-2U25), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18430986 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430986 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430986 Supplier Novus Biologicals Supplier No. NBP212158AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DC-SIGNR/CD299/CLEC4M Monoclonal antibody specifically detects DC-SIGNR/CD299/CLEC4M in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen DC-SIGNR/CD299/CLEC4M
Applications Flow Cytometry, Immunohistochemistry, CyTOF, Neutralization
Classification Monoclonal
Clone MM0241-2U25
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias CD209L1, CD209Ldendritic cell-specific ICAM-3-grabbing nonintegrin 2, CD299, CD299 antigenMGC129964, C-type lectin domain family 4, member M, DC-SIGN2CD209 antigen-like protein 1, DC-SIGNRDC-SIGN-related protein, DCSIGNRMGC47866, Dendritic cell-specific ICAM-3-grabbing non-integrin 2, HP10347, liver/lymph node-specific ICAM-3 grabbing non-integrin, Liver/lymph node-specific ICAM-3-grabbing non-integrin, L-SIGN, LSIGNC-type lectin domain family 4 member M, mannose binding C-type lectin DC-SIGNR
Host Species Mouse
Immunogen Human recombinant DC-SIGNR/CD299/CLEC4M
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Signal Transduction, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 10332
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.