Learn More
DC-SIGNR/CD299/CLEC4M Antibody (16E7), DyLight 680 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52649FR
417.35 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18759692
  • £486.00 / 0.10mL
Description
DC-SIGNR/CD299/CLEC4M Monoclonal antibody specifically detects DC-SIGNR/CD299/CLEC4M in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DC-SIGNR/CD299/CLEC4M
Monoclonal
DyLight 680
CD209L1, CD209Ldendritic cell-specific ICAM-3-grabbing nonintegrin 2, CD299, CD299 antigenMGC129964, C-type lectin domain family 4, member M, DC-SIGN2CD209 antigen-like protein 1, DC-SIGNRDC-SIGN-related protein, DCSIGNRMGC47866, Dendritic cell-specific ICAM-3-grabbing non-integrin 2, HP10347, liver/lymph node-specific ICAM-3 grabbing non-integrin, Liver/lymph node-specific ICAM-3-grabbing non-integrin, L-SIGN, LSIGNC-type lectin domain family 4 member M, mannose binding C-type lectin DC-SIGNR
CD299
0.1 mL
Signal Transduction
10332
Store at 4°C in the dark.
IgG κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, CyTOF
16E7
50 mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified