missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DC-SIGNR/CD299/CLEC4M Antibody (120604) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB162JF585
This item is not returnable.
View return policy
Description
DC-SIGNR/CD299/CLEC4M Monoclonal antibody specifically detects DC-SIGNR/CD299/CLEC4M in Human samples. It is validated for Flow Cytometry,Adhesion Blockade,CyTOF
Specifications
| DC-SIGNR/CD299/CLEC4M | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CD209L1, CD209Ldendritic cell-specific ICAM-3-grabbing nonintegrin 2, CD299, CD299 antigenMGC129964, C-type lectin domain family 4, member M, DC-SIGN2CD209 antigen-like protein 1, DC-SIGNRDC-SIGN-related protein, DCSIGNRMGC47866, Dendritic cell-specific ICAM-3-grabbing non-integrin 2, HP10347, liver/lymph node-specific ICAM-3 grabbing non-integrin, Liver/lymph node-specific ICAM-3-grabbing non-integrin, L-SIGN, LSIGNC-type lectin domain family 4 member M, mannose binding C-type lectin DC-SIGNR | |
| NIH-3T3 mouse embryonic fibroblast cell line transfected with human DC-SIGNR/CD299, Accession # Q9H2X3 | |
| 0.5 mL | |
| Adaptive Immunity, Immunology, Signal Transduction, Virology Bacteria and Parasites | |
| 10332 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Adhesion Blockade, CyTOF | |
| 120604 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction