missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DC-SIGN+DC-SIGNR Antibody (DC28), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB16211A
This item is not returnable.
View return policy
Description
DC-SIGN+DC-SIGNR Monoclonal antibody specifically detects DC-SIGN+DC-SIGNR in Human samples. It is validated for Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF
Specifications
| DC-SIGN+DC-SIGNR | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 30835 | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF | |
| DC28 | |
| Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF-ready | |
| CD209,CD209 antigen,CDSIGN,CLEC4L,DC-SIGN,DC-SIGN1,hDC-SIGN | |
| E. coli-derived recombinant human DC-SIGN, Extracellular domain | |
| 0.5 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction