missing translation for 'onlineSavingsMsg'
Learn More

DC-SIGN+DC-SIGNR Antibody (120612) [PerCP], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals FAB1621C

439.45 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


Product Code. 18646443

  • £550.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DC-SIGN+DC-SIGNR Monoclonal antibody specifically detects DC-SIGN+DC-SIGNR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DC-SIGN+DC-SIGNR
Monoclonal
PerCP
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
30835
Store at 4°C in the dark.
IgG2a
Flow Cytometry, Immunohistochemistry, CyTOF
120612
Flow Cytometry, Immunohistochemistry, CyTOF-ready
CD209;CD209 antigen;CDSIGN;CLEC4L;DC-SIGN;DC-SIGN1;hDC-SIGN
NIH-3T3 mouse embryonic fibroblast cell line transfected with human DC-SIGNR, Accession # Q9H2X3
0.1 mL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.